missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR125 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518934
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518934 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518934 Supplier Invitrogen™ Supplier No. PA518934
Only null left

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR125 (G protein-coupled receptor 125), also known as PGR21 or TEM5L, is a 1,321 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 2 family and the LN-TM7 subfamily. Considered a novel orphan adhesion-type G-protein-coupled receptor, GPR125 has five leucine rich repeats (LRR), an immunoglobulin (Ig) domain and a GPS domain. GPR125 may play a functional role in choroidal and hippocampal response to brain injury. It is also suggested that GPR125 may be a marker for spermatogonial stem cells. Four isoforms of GPR125 exists due to alternative splicing events.

Specifications

Antigen GPR125
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ADGRA3
Gene Accession No. Q8IWK6
Gene Alias 3830613O22Rik; Adgra2; ADGRA3; adhesion G protein-coupled receptor A2; adhesion G protein-coupled receptor A3; AU044632; G protein-coupled receptor 125; Gpr125; G-protein coupled receptor 125; PGR21; probable G-protein coupled receptor 125; TEM5L; Tem5-like; UNQ556/PRO1113
Gene Symbols ADGRA3
Host Species Goat
Immunogen Synthetic peptide sequence (CTNKSASSFKNS) corresponding to the internal amino acids of GPR125 (aa 1098-1110).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166647
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less