missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP9, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002815B01P
Description
Sequence: MPAWGALFLLWATAEATKDCPSPCTCRALETMGLWVDCRGHGLTALPALPARTRHLLLANNSLQSVPPGAFDHLPQLQTLDVTQNPWHCDCSLTYLRLWLEDRTPEALLQVRCASPSLAAHGPLGRLTGYQLGSCGWQLQASWVRPGVLWDVALVTVAALGLALLAGLLCATTEALDSpecifications
| GP9 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC030229.1 | |
| GP9 | |
| GP9 (AAH30229.1, 1 a.a. ∼ 177 a.a) full-length human protein. | |
| RUO | |
| 2815 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| glycoprotein IX (platelet) | |
| CD42a/GPIX | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |