Learn More
Invitrogen™ Mouse gp130 (IL 6ST) ELISA Kit
Description
The Mouse gp130 (IL-6ST) ELISA quantitates Ms gp130 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms gp130. Principle of the method The Mouse gp130 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | Q00560 |
| Assay Range | 40.96 to 10,000 pg/mL |
| Assay Sensitivity | 40 pg/mL |
| Calibration Range | 40 pg/mL to 10000 pg/mL |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Product Type | ELISA |
| Protein Family | Cytokines & Receptors |
| Sample Type | Plasma, Serum, Supernatant |
| Specificity | This ELISA antibody pair detects human mouse gp-130. Other species not determined yet |
| Show More |