missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GoTaq™ G2 DNA Polymerase
GoTaq™ G2 DNA Polymerase is supplied with 5X Green GoTaq™ Reaction Buffer and 5X Colorless GoTaq™ Reaction Buffer.
Specifications
| Concentration | 5 U/μL |
|---|---|
| Content And Storage | −30° to −10°C |
| Description | DNA Polymerase |
| For Use With (Application) | RT-PCR |
Description
- Next-Generation GoTaq(R) for Robust, Reliable PCR Amplification
- Full-length Taq DNA polymerase that exhibits 5′→3′ exonuclease activity
- Direct-to-gel convenience
- 5X Reaction Buffer with MgCl2 to simplify reaction setup
- Supplied at a concentration of 5u/μL
Specifications
| 5 U/μL | |
| −30° to −10°C | |
| DNA Polymerase | |
| RT-PCR |