Learn More
Promega GoTaq™ G2 DNA Polymerase
GoTaq™ G2 DNA Polymerase is supplied with 5X Green GoTaq™ Reaction Buffer and 5X Colorless GoTaq™ Reaction Buffer.
Supplier:  Promega M7845
Encompass
 View more versions of this product
Catalog No. PRM7845
Only null left
Description
Description
  • Next-Generation GoTaq(R) for Robust, Reliable PCR Amplification
  • Full-length Taq DNA polymerase that exhibits 5′→3′ exonuclease activity
  • Direct-to-gel convenience
  • 5X Reaction Buffer with MgCl2 to simplify reaction setup
  • Supplied at a concentration of 5u/μL
Specifications
Chemical Identifiers
5 U/μL
−30° to −10°C
DNA Polymerase
RT-PCR
Specifications
5 U/μL
DNA Polymerase
RT-PCR
−30° to −10°C
500U
Product Suggestions
Product Suggestions