missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GOLIM4 Recombinant Rabbit Monoclonal Antibody (23GB5575)
GREENER_CHOICE

Catalog No. MA553900
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553900 100 μL
MA553901 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553900 Supplier Invitrogen™ Supplier No. MA553900
Only null left

Rabbit Recombinant Monoclonal Antibody

GOLIM4 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi-resident protein. It may process proteins synthesized in the rough endoplasmic reticulum and assist in the transport of protein cargo through the Golgi apparatus.

Specifications

Antigen GOLIM4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5575
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GOLIM4
Gene Accession No. O00461, Q5BJK8, Q8BXA1
Gene Alias 130 kDa golgi-localized phosphoprotein; 3110027H23Rik; cis Golgi-localized calcium-binding protein; decapacitation factor 10; Df10; GIMPc; Golgi integral membrane protein 4; Golgi integral membrane protein, cis; golgi phosphoprotein 4; Golgi phosphoprotein of 130 kDa; golgi-localized phosphoprotein of 130 kDa; GOLIM4; GOLPH4; GPP130; LOW QUALITY PROTEIN: Golgi integral membrane protein 4; P138; type II Golgi membrane protein
Gene Symbols GOLIM4
Host Species Rabbit
Immunogen A synthesized peptide derived from human GOLPH4 (50-100AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27333, 310526, 73124
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less