missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Golgi protein 58k Recombinant Rabbit Monoclonal Antibody (23GB5765)

Rabbit Recombinant Monoclonal Antibody

$590.92 - $1416.21

Specifications

Antigen Golgi protein 58k
Clone 23GB5765
Concentration 0.75 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA552912
View Documents
Invitrogen™
MA552912
100 μL
Each for $1,416.21
Only null left
 
MA552913
View Documents
Invitrogen™
MA552913
20 μL
Each for $590.92
Only null left
 
Description

Description

Golgi protein 58k Recombinant Monoclonal Antibody for Western Blot, Flow

Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.
Specifications

Specifications

Golgi protein 58k
0.75 mg/mL
Flow Cytometry, Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95954
10841
A synthesized peptide derived from human 58K Golgi protein (50-100AA).
Antibody
23GB5765
-20°C
Recombinant Monoclonal
Liquid
IgG
Human
Ftcd
58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1
Ftcd
Primary
Affinity chromatography
Videos
SDS
Documents

Documents