missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Golgi protein 58k Recombinant Rabbit Monoclonal Antibody (23GB5765)
Description
Golgi protein 58k Recombinant Monoclonal Antibody for Western Blot, Flow
Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
| Antigen | Golgi protein 58k |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 23GB5765 |
| Concentration | 0.75 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Ftcd |
| Gene Accession No. | O95954 |
| Gene Alias | 58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1 |
| Show More |