missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate, Monohydrate, Powder, FCC, BP, 98-102%, Spectrum™ Chemical

Catalog No. 18604207
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604207 50 kg Poly Bottle
18604204 500 g Poly Bottle
18604205 2.5 kg Poly Bottle
18604206 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604207 Supplier Spectrum Chemical Mfg Cor Supplier No. MA16450KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MA164, 10034-96-5, MnSO4·H2O

Spectrum™ Chemical Manganese Sulfate, Monohydrate, Powder, FCC, BP - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Physical Form Prills or Crystals
CAS Max % 100%
Quantity 50 kg
Packaging Poly Bottle
Percent Purity 98 to 102%
Grade BP/FCC
Chemical Name or Material Manganese Sulfate, Monohydrate, Powder