missing translation for 'onlineSavingsMsg'
Learn More

Gold wire, 2.0mm (0.08in) dia, Premion™, 99.999% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol

$328.34 - $6129.14

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA14733BO
View Documents
Thermo Scientific Chemicals
014733BO
1 cm
Each for $328.34
Only null left
 
AA14733BQ
View Documents
Thermo Scientific Chemicals
014733BQ
5 cm
Each for $1,636.44
Only null left
 
AA14733BU
View Documents
Thermo Scientific Chemicals
014733BU
25 cm
Each for $6,129.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]

Specifications

7440-57-5
Odorless
Wire
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
99.999%
Gold wire, 2.0mm (0.08 in.) dia.
1 cm
≈60.6g/m
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
196.97
CHEBI:30050
Premion
(metals basis)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-165-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only