missing translation for 'onlineSavingsMsg'
Learn More

Gold wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis)

Catalog No. p-4824735
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 cm
2 cm
50 cm
Catalog No. Quantity
AA14726BP 2 cm
AA14726BS 10 cm
AA14726BW 50 cm
3 options

Catalog No. AA14726BP

Supplier: Thermo Scientific Chemicals 014726BP

Only null left

Form: Approx.15.15g/m | CAS: 7440-57-5 | Au | 196.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Golden
Appearance Gold colored wire
Quantity 2 cm
Odor Odorless
Form Wire
Assay Percent Range 99.999% (Metals basis)
Chemical Name or Material Gold
EINECSNumber 231-165-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only