missing translation for 'onlineSavingsMsg'
Learn More

Gold wire, 1.0mm (0.04in) dia, 99.95% (metals basis)

Catalog No. p-7049569
Change view
Click to view available options
Quantity:
10 cm
2 cm
50 cm
Catalog No. Quantity
AA14731BP 2 cm
AA14731BS 10 cm
AA14731BW 50 cm
3 options

Catalog No. AA14731BP

Supplier: Thermo Scientific Chemicals 014731BP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-57-5 | Au | 196.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Quantity 2 cm
Odor Odorless
Weight ≈15.15g/m
Form Wire
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Gold wire, 1.0mm (0.04 in.) dia.
EINECSNumber 231-165-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only