missing translation for 'onlineSavingsMsg'
Learn More

Gold wire, 0.25mm (0.01in) dia, Premion™, 99.998% (metals basis)

Catalog No. AA10967BU
Change view
Click to view available options
Quantity:
25 cm
100 cm
500 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10967BU 25 cm
AA10967BY 100 cm
AA10967CC 500 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10967BU Supplier Thermo Scientific Chemicals Supplier No. 010967BU

CAS: 7440-57-5 | Au | 196.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Quantity 25 cm
Odor Odorless
Weight ≈0.95g/m
Form Wire
Percent Purity 99.998%
Grade Premion
Assay (metals basis)
Chemical Name or Material Gold wire, 0.25mm (0.01 in.) dia.
EINECSNumber 231-165-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only