missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold Ultrathin foil, 1.0 micron thick, 99.9% (metals basis), Thermo Scientific Chemicals
$812.04 - $812.04
Chemical Identifiers
| CAS | 7440-57-5 |
|---|---|
| Molecular Formula | Au |
| Molecular Weight (g/mol) | 196.97 |
| MDL Number | MFCD00003436 |
| InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
| Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
| PubChem CID | 23985 |
| ChEBI | CHEBI:30050 |
| IUPAC Name | gold |
| SMILES | [Au] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-57-5 | |
| 196.97 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| 23985 | |
| gold |
| Au | |
| MFCD00003436 | |
| colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
| CHEBI:30050 | |
| [Au] |
Specifications
| 7440-57-5 | |
| Odorless | |
| MFCD00003436 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| gold | |
| 23985 | |
| 99.9% | |
| (metals basis) |
| 25 x 25 mm | |
| Au | |
| colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
| [Au] | |
| 196.97 | |
| CHEBI:30050 | |
| Gold Ultrathin foil, 1.0μm thick |
Safety and Handling
EINECSNumber : 231-165-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only