missing translation for 'onlineSavingsMsg'
Learn More

Gold Thinfoil, 0.01mm (0.0004in) thick, 99.9% (metals basis)

Catalog No. AA11518FF
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11518FF 25 x 25 mm
AA11518FI 50 x 50 mm
AA11518GH 100 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA11518FF Supplier Thermo Scientific Chemicals Supplier No. 011518FF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-57-5 | Au | 196.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Quantity 25 x 25 mm
Odor Odorless
Weight ≈0.12g/25x25mm
Form Foil
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Gold Thinfoil, 0.01mm (0.0004 in.) thick, not certified pinhole free
EINECSNumber 231-165-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only