missing translation for 'onlineSavingsMsg'
Learn More

Gold slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Premion™, 99.995% (metals basis)

Catalog No. p-7051094
Change view
Click to view available options
Quantity:
1 Pc.
5 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40442KG 1 Pc.
AA40442KJ 5 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40442KG Supplier Thermo Scientific Chemicals Supplier No. 040442KG
Only null left

Electronics, vapor deposition | CAS: 7440-57-5 | Au | 196.97 g/mol

Electronics, vapor depositionGold slug is used in thermal evaporation and electrodes (quartz oscillator, light-emitting diodes, and laser diodes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Quantity 1 Pc.
Odor Odorless
Weight ≈0.49g/slug
Form Slug
Solubility Information Soluble in hot sulfuric acid and aqua regia. Insoluble in water and acid.
Percent Purity 99.995%
Grade Premion
Assay (metals basis)
Chemical Name or Material Gold slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length
EINECSNumber 231-165-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only