missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold powder, spherical, APS 0.5-0.8 micron, 99.96+% (metals basis)
Form: S.A. 0.45-0.70 m2/g | CAS: 7440-57-5 | Au | 196.97 g/mol
$152.53 - $2527.92
Chemical Identifiers
| CAS | 7440-57-5 |
|---|---|
| Molecular Formula | Au |
| Molecular Weight (g/mol) | 196.97 |
| MDL Number | MFCD00003436 |
| InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
| Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
| PubChem CID | 23985 |
| ChEBI | CHEBI:30050 |
| IUPAC Name | gold |
| SMILES | [Au] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA44636MD
|
Thermo Scientific Chemicals
44636MD |
250 mg |
Each for $152.53
|
|
|||||
|
AA4463603
|
Thermo Scientific Chemicals
04463603 |
1 g |
Each for $531.36
|
|
|||||
|
AA4463606
|
Thermo Scientific Chemicals
04463606 |
5 g |
Each for $2,527.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-57-5 | |
| 196.97 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| 23985 | |
| gold |
| Au | |
| MFCD00003436 | |
| colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
| CHEBI:30050 | |
| [Au] |
Specifications
| Golden | |
| Gold colored powder | |
| Odorless | |
| Spherical Powder | |
| MFCD00003436 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| gold | |
| 23985 | |
| Gold |
| 7440-57-5 | |
| 250 mg | |
| >99.96% (Metals basis) | |
| Au | |
| colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
| [Au] | |
| 196.97 | |
| CHEBI:30050 |
Safety and Handling
EINECSNumber : 231-165-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only