missing translation for 'onlineSavingsMsg'
Learn More

Gold powder, -100 mesh, Premion™, 99.998% (metals basis)

Catalog No. p-4781909
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4488203 1 g
AA4488206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4488203 Supplier Thermo Scientific Chemicals Supplier No. 04488203
Only null left

Gold, Au, CAS Number-7440-57-5, burnish, leaf, c.i. pigment metal 3, flake, magnesium purple, colloidal, ci pigment metal 3, shell, powder, gold, colloidal, 5g, Gold colored powder, 99.998% (Metals basis), CHEBI:30050, Golden, 19.32g/cm3 at 20 deg.C, 231-165-9, PCHJSUWPFVWCPO-UHFFFAOYSA-N, -100 Mesh | CAS: 7440-57-5 | Au | 196.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Golden
Appearance Gold colored powder
Quantity 1 g
Mesh Size -100 Mesh
Odor Odorless
Form Powder
Assay Percent Range 99.998% (Metals basis)
Chemical Name or Material Gold
EINECSNumber 231-165-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only