missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold plating solution, electroless, metal content≈3.7g/L
Description
For coating nickel or copper
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Gold plating solution |
| Color | Colorless to Yellow |
| Name Note | Electroless, metal content ≈3.7g/L |
| MDL Number | MFCD00003436 |
| UN Number | UN2922 |
| Quantity | 250 mL |
| Physical Form | Liquid |
RUO – Research Use Only