Learn More
Gold nanoparticles, 5nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 515-520nm absorption
CAS: 7440-57-5 | Au | 196.97 g/mol
$217.14 - $683.18
Chemical Identifiers
| CAS | 7440-57-5 |
|---|---|
| Molecular Formula | Au |
| Molecular Weight (g/mol) | 196.97 |
| MDL Number | MFCD00003436 |
| InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
| Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
| PubChem CID | 23985 |
| ChEBI | CHEBI:30050 |
| IUPAC Name | gold |
| SMILES | [Au] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ67448AC
|
Thermo Scientific Chemicals
J67448AC |
25 mL |
Each for $217.14
|
|
|||||
|
AAJ67448AE
|
Thermo Scientific Chemicals
J67448AE |
100 mL |
Each for $683.18
|
|
|||||
Description
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-57-5 | |
| 196.97 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| 23985 | |
| gold |
| Au | |
| MFCD00003436 | |
| colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
| CHEBI:30050 | |
| [Au] |
Specifications
| Red | |
| 25 mL | |
| 5nm | |
| Liquid | |
| MFCD00003436 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| gold | |
| 23985 | |
| 99% |
| 7440-57-5 | |
| Odorless | |
| Light sensitive | |
| Au | |
| colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
| [Au] | |
| 196.97 | |
| CHEBI:30050 | |
| Gold nanoparticles, supplied in 0.1mM PBS, reactant to free, 99%, OD1, 515 to 520nm absorption |
Safety and Handling
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only