missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold nanoparticles, 400nm, supplied in 0.1mM PBS, 95%, Thermo Scientific Chemicals
$340.70 - $340.70
Chemical Identifiers
| CAS | 7440-57-5 |
|---|---|
| Molecular Formula | Au |
| Molecular Weight (g/mol) | 196.97 |
| MDL Number | MFCD00003436 |
| InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
| Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
| PubChem CID | 23985 |
| ChEBI | CHEBI:30050 |
| IUPAC Name | gold |
| SMILES | [Au] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ67106AC
|
Thermo Scientific Chemicals
J67106AC |
25 mL |
Each for $340.70
|
|
|||||
Description
For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
Chemical Identifiers
| 7440-57-5 | |
| 196.97 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| 23985 | |
| gold |
| Au | |
| MFCD00003436 | |
| colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
| CHEBI:30050 | |
| [Au] |
Specifications
| Pink to Red | |
| 25 mL | |
| Light sensitive | |
| Au | |
| colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
| [Au] | |
| 196.97 | |
| CHEBI:30050 | |
| Gold nanoparticles, supplied in 0.1mM PBS |
| 7440-57-5 | |
| 400nm | |
| Liquid | |
| MFCD00003436 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| gold | |
| 23985 | |
| 95% |
Safety and Handling
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only