missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 20nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 524nm absorption

CAS: 7440-57-5 | Au | 196.97 g/mol

$238.32 - $674.34

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67370AC
View Documents
Thermo Scientific Chemicals
J67370AC
25 mL
Each for $238.32
Only null left
 
AAJ67370AE
View Documents
Thermo Scientific Chemicals
J67370AE
100 mL
Each for $674.34
Only null left
 
Description

Description

Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]

Specifications

Pink to Red
25 mL
20nm
Liquid
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
99%
7440-57-5
Odorless
Light sensitive
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
196.97
CHEBI:30050
Gold nanoparticles, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 524nm absorption
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only