missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 20nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 524nm absorption

Catalog No. p-7057749
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67269AC 25 mL
AAJ67269AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67269AC Supplier Thermo Scientific Chemicals Supplier No. J67269AC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-57-5 | Au | 196.97 g/mol

Colloidal gold is an electron-dense, non-fading marker useful as a probe in electron microscopy, light microscopy and immunoblotting. It requires no additional processing for detection, but in some applications the signal can be enhanced by reaction with silver. It can be complexed with biomolecules by strong, non-covalent interactions. Particles less than 15 nm are most useful in transmission electron microscopy and for applications where access to the probe may be hindered by larger particles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Colloidal gold is an electron-dense, non-fading marker useful as a probe in electron microscopy, light microscopy and immunoblotting. It requires no additional processing for detection, but in some applications the signal can be enhanced by reaction with silver. It can be complexed with biomolecules by strong, non-covalent interactions. Particles less than 15 nm are most useful in transmission electron microscopy and for applications where access to the probe may be hindered by larger particles.

Solubility
Fully miscible in water. It is soluble in aqua regia, nitric acid and hydrochloric acid.

Notes
Particles per ml: 6.54E+11

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Pink to Red
Quantity 25 mL
Particle Size 20nm
Sensitivity Light sensitive
Form Liquid
Solubility Information Fully miscible in water. It is soluble in aqua regia,nitric acid and hydrochloric acid.
Chemical Name or Material Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 524nm absorption
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only