missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 15nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption

Catalog No. p-7057761
Change view
Click to view available options
Quantity:
100 mL
25 mL
Catalog No. Quantity
AAJ67286AC 25 mL
AAJ67286AE 100 mL
2 options

Catalog No. AAJ67286AC

Supplier: Thermo Scientific Chemicals J67286AC

Only null left

CAS: 7440-57-5 | Au | 196.97 g/mol

Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Red
Quantity 25 mL
Particle Size 15nm
Sensitivity Light sensitive
Form Liquid
Percent Purity 99%
Chemical Name or Material Gold nanoparticles, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only