missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 10nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 520nm absorption

Catalog No. p-7057703
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67188AC 25 mL
AAJ67188AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67188AC Supplier Thermo Scientific Chemicals Supplier No. J67188AC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-57-5 | Au | 196.97 g/mol

For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Pink to Red
Quantity 25 mL
Odor Odorless
Particle Size 10nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 520nm absorption
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only