Learn More
Thermo Scientific™ GC Injection Port Base Seals for Thermo Scientific™ and Agilent Instruments
Ensure optimal GC analysis by using the exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals.
$160.90 - $954.49
Products 8
Catalog Number Mfr. No. Diameter (Metric) Inner For Use With (Equipment) Quantity Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner For Use With (Equipment) Quantity Product Type Price Quantity & Availability  
03150333
View Documents Promotion Details
Thermo Scientific™
290GA081
0.8 mm Single Column Installation 10 Pack Gold Inlet Base Seal
Pack of 10 Each for $863.05
Only null left
03150332
View Documents Promotion Details
Thermo Scientific™
290GA082
0.8 mm Single Column Installation 2 Pack Gold Inlet Base Seal
Pack of 2 Each for $263.77
Only null left
03150330
View Documents Promotion Details
Thermo Scientific™
290GA122
1.2 mm Dual Column Installation 2 Pack Gold Inlet Base Seal
Pack of 2 Each for $213.49
Only null left
03150380
View Documents Promotion Details
Thermo Scientific™
290GA083
0.8 mm Single Column Installation (Cross Version) 2 Pack Gold Inlet Base Seal
Pack of 2 Each for $190.90
Only null left
03150381
View Documents Promotion Details
Thermo Scientific™
290GA084
0.8 mm Single Column Installation (Cross Version) 10 Pack Gold Inlet Base Seal
Pack of 10 Each for $861.59
Only null left
03150376
View Documents Promotion Details
Thermo Scientific™
290GA091
0.8 mm Single Column Installation 10 Pack Siltek Inlet Base Seal
Pack of 10 Each for $725.41
Only null left
03150377
View Documents Promotion Details
Thermo Scientific™
290GA092
0.8 mm Single Column Installation 2 Pack Siltek Inlet Base Seal
Pack of 2 Each for $160.90
Only null left
03150379
View Documents Promotion Details
Thermo Scientific™
290GA094
0.8 mm Single Column Installation (Cross Version) 10 Pack Siltek Inlet Base Seal
Pack of 10 Each for $954.49
Only null left
Description
Description
Ensure optimal gas chromatography (GC) analysis by using exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals. Precision machined from high grade stainless steel, choose from gold-plated or Siltek-treated base seals. The chemically inert coated surface makes them suitable for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenols. Use these GC Injection Port Base Seals with split/splitless injection ports of thermo Scientific™ TRACE™ 1300 Series GC and Agilent GC 5890/6890/7890 systems.
  • Precision machined to provide exceptional sealing properties
  • Reduces activity (especially useful for analysis of pesticide, PCBs, phenols, etc.)
  • High grade stainless steel provides reproducible seal
  • Direct equivalent to Agilent original part
  • Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports
Videos
SDS
Documents
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.