missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride, Au 64.4% min

Catalog No. p-3257007
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1216301 0.25 g
AA1216303 1 g
AA1216306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1216301 Supplier Thermo Scientific Chemicals Supplier No. 01216301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13453-07-1 | AuCl3

It is used in colloidal gold solutions, in photography and as a print toning agent(gold toning), starting solution to form other gold compounds and a precursor for preparation of ultra pure gold metal. It is used in electroplating and electroless plating as an anode in an electric cell. Gold(III) chloride acts as a gold catalyst and cell body stains for bright field and dark field microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13453-07-1
Molecular Formula AuCl3
MDL Number MFCD00014172

Specifications

Melting Point 180° (sublimation)
Color Red
Physical Form Crystalline
Quantity 0.25 g
Solubility Information Soluble in water,ethanol,ether and slightly soluble in liquid ammonia.
Formula Weight 303.33
Odor Odorless
Sensitivity Hygroscopic
Appearance Red crystalline
Density 3.9
Chemical Name or Material Gold(III) chloride
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation DOT Class: 8, Packing Group: III
EINECSNumber 236-623-1
RTECSNumber MD5420000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only