Learn More
Gold(III) chloride, 99%
CAS: 13453-07-1 | AuCl3 | 303.32 g/mol
Supplier: Thermo Scientific Chemicals 318640050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Gold(III) chloride | |
| 99% | |
| 13453-07-1 | |
| 5 g | |
| AuCl3 | |
| MFCD00014172 | |
| Solubility in water: soluble | |
| 303.32 | |
| Glass bottle |
| 254.0°C | |
| Brown to Yellow | |
| Crystalline Powder, Crystals and/or Chunks | |
| 98.5% min. (ex Au) | |
| AuCl3 | |
| 15, 4556 | |
| 303.32 | |
| 99% |
Chemical Identifiers
| 13453-07-1 | |
| AuCl3 | |
| 303.32 | |
| MFCD00014172 |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 236-623-1
RTECSNumber : MD5420000
TSCA : TSCA
RUO – Research Use Only