missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride, 99.99+%, (trace metal basis)

Catalog No. AC437080050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437080050 5 g Glass bottle
AC437080010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437080050 Supplier Thermo Scientific Chemicals Supplier No. 437080050

CAS: 13453-07-1 | AuCl3 | 303.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 13453-07-1
Molecular Formula AuCl3
Molecular Weight (g/mol) 303.32

Specifications

Melting Point 254.0°C
Color Brown to Red
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Assay Percent Range 63.0 to 66.9% (Au)
Linear Formula AuCl3
Merck Index 15,4556
Solubility Information Solubility in water: soluble
Formula Weight 303.32
Percent Purity ≥99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gold(III) chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 236-623-1

RUO – Research Use Only