missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) acetate, 99.9% (metals basis)

Catalog No. AA3974202
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3974202 0.5 g
AA3974203 1 g
AA3974206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3974202 Supplier Thermo Scientific Chemicals Supplier No. 03974202

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15804-32-7 | C6H9AuO6 | 374.099 g/mol

Used in solar energy technologies. Used in petrochemical cracking and automotive catalysts, water treatment, plating, textiles, research and in optic, laser, crystal and glass applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15804-32-7
Molecular Formula C6H9AuO6
Molecular Weight (g/mol) 374.099
MDL Number MFCD00210614
InChI Key OTCKNHQTLOBDDD-UHFFFAOYSA-K
Synonym gold 3+ triacetate, gold iii acetate, gold acetate, acmc-1c3i2, triacetic acid gold iii salt, acetic acid, gold 3+ salt 3:1
PubChem CID 15952561
IUPAC Name gold(3+);triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Au+3]

Specifications

Color Brown
Physical Form Powder
Quantity 0.5 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Slightly soluble in water.
Formula Weight 374.1
Appearance Brown powder
Chemical Name or Material Gold(III) acetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only