Learn More
Gold(I) potassium cyanide, Premion™, 99.96% (metals basis), Au 67.6% min
CAS: 13967-50-5
$476.63 - $2106.01
Chemical Identifiers
| CAS | 13967-50-5 |
|---|---|
| MDL Number | MFCD00011414 |
| Synonym | Potassium dicyanoaurate(I); Potassium gold cyanide |
Description
Gold potassium cyanide is an important gold source used in electrolytic gold plating. The primary application of gold potassium cyanide is in the electroplating of metals. In addition, gold potassium cyanide is also used in the decorative plating of jewelry. It is useful for photoreduction of gold ions using the ZnO nanopowder. It is useful for the preparation of gold-gold junction electrode in the voltammetric glucose detection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13967-50-5 | |
| MFCD00011414 | |
| Potassium dicyanoaurate(I); Potassium gold cyanide |
Specifications
| (decomposition) | |
| White | |
| 1 g | |
| MFCD00011414 | |
| 6235525 | |
| Potassium dicyanoaurate(I); Potassium gold cyanide | |
| 288.33 | |
| Premion | |
| Odorless | |
| Gold(I) potassium cyanide |
| 13967-50-5 | |
| Crystalline Powder | |
| KAu(CN)2 | |
| UN1588 | |
| 14,7628 | |
| Soluble in water and slightly soluble in alcohol. | |
| 99.96% | |
| Au 67.6% min. | |
| 3.45 g/mL | |
| (metals basis) |
Safety and Handling
GHS H Statement
H300-H310-H330-H318-H290-H315
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.
P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P330-P332+P313-P361-P363-P390-P501c
H290-H300+H310+H330-H315-H318
DOTInformation : Hazard Class: 6.1; Packaging Group: I
EINECSNumber : 237-748-4
RTECSNumber : CK6270000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only