accessibility menu, dialog, popup

UserName

KAuCNCN

CAS RN 13967-50-5

IUPAC Name: gold(1+) potassium bis(iminomethanide)
Synonyms: gold(1+) potassium bis(cyanide)
Molecular Weight (g/mol): 288.1
Molecular Formula: C2AuKN2
InChi Key: XTFKWYDMKGAZKK-UHFFFAOYSA-N
SMILES: [K+].[Au+].[C-]#N.[C-]#N
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)