Learn More
Gold(I) chloride, Premion™, 99.99% (metals basis), Au 84.2% min
CAS: 10294-29-8 | AuCl | 232.42 g/mol
$171.23 - $2321.37
Chemical Identifiers
| CAS | 10294-29-8 |
|---|---|
| Molecular Formula | AuCl |
| Molecular Weight (g/mol) | 232.42 |
| MDL Number | MFCD00046175 |
| InChI Key | FDWREHZXQUYJFJ-UHFFFAOYSA-M |
| Synonym | gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade |
| PubChem CID | 27366 |
| ChEBI | CHEBI:30078 |
| IUPAC Name | chlorogold |
| SMILES | [Cl-].[Au+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4043201
|
Thermo Scientific Chemicals
04043201 |
0.25 g |
Each for $171.23
|
|
|||||
|
AA4043203
|
Thermo Scientific Chemicals
04043203 |
1 g |
Each for $534.89
|
|
|||||
|
AA4043206
|
Thermo Scientific Chemicals
04043206 |
5 g |
Each for $2,321.37
|
|
|||||
Description
As intermediates. Also In Chemicals, pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10294-29-8 | |
| 232.42 | |
| FDWREHZXQUYJFJ-UHFFFAOYSA-M | |
| 27366 | |
| chlorogold |
| AuCl | |
| MFCD00046175 | |
| gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade | |
| CHEBI:30078 | |
| [Cl-].[Au+] |
Specifications
| 289°C (decomposition) | |
| Powder | |
| AuCl | |
| UN3260 | |
| gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade | |
| FDWREHZXQUYJFJ-UHFFFAOYSA-M | |
| chlorogold | |
| 27366 | |
| 232.42 | |
| Premion | |
| Odorless | |
| 7.57 g/mL | |
| (metals basis) |
| 10294-29-8 | |
| 0.25 g | |
| MFCD00046175 | |
| 14,4515 | |
| Very slightly soluble in water. soluble in HCl,HBr organic solvents. | |
| [Cl-].[Au+] | |
| 232.42 | |
| CHEBI:30078 | |
| 99.99% | |
| Au 84.2% min. | |
| Moisture sensitive | |
| Gold(I) chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P272-P280-P301+P330+P331-P302+P352-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H317-H500
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 233-655-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only