missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) chloride, Premion™, 99.99% (metals basis), Au 84.2% min

CAS: 10294-29-8 | AuCl | 232.42 g/mol

Supplier:  Thermo Scientific Chemicals 04043203

 View more versions of this product

Catalog No. AA4043203


Only null left
Explore available promotions

Description

Description

As intermediates. Also In Chemicals, pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gold(I) chloride
10294-29-8
1 g
AuCl
UN3260
gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade
FDWREHZXQUYJFJ-UHFFFAOYSA-M
chlorogold
27366
232.42
Premion
Odorless
7.57 g/mL
289°C (decomposition)
Powder
(metals basis)
MFCD00046175
14,4515
Very slightly soluble in water. soluble in HCl,HBr organic solvents.
[Cl-].[Au+]
232.42
CHEBI:30078
99.99%
Au 84.2% min.
Moisture sensitive

Chemical Identifiers

10294-29-8
232.42
FDWREHZXQUYJFJ-UHFFFAOYSA-M
27366
chlorogold
AuCl
MFCD00046175
gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade
CHEBI:30078
[Cl-].[Au+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P234-P260-P264b-P272-P280-P301+P330+P331-P302+P352-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H317-H500

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 233-655-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only