missing translation for 'onlineSavingsMsg'
Learn More

Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis)

Form: Wire cloth; Open area: 62.7%; Width of opening: 0.015 in. | CAS: 7440-57-5 | Au | 196.97 g/mol

Supplier:  Thermo Scientific Chemicals 040930FY

 View more versions of this product

Catalog No. AA40930FY


Only null left
Explore available promotions

Description

Description

Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis) is used as an electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Golden
7440-57-5
50 x 100 mm
Odorless
Wire Cloth
MFCD00003436
Soluble in aqua regia.
[Au]
196.97
CHEBI:30050
Gold
Gold colored gauze
52 Mesh
99.99% (Metals basis)
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-165-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only