Learn More
Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis)
Form: Wire cloth 100 x 100mm Approx.4.9g; Open area: 56.3%; Width of opening: 0.0075 in. | CAS: 7440-57-5 | Au | 196.97 g/mol
Supplier: Thermo Scientific Chemicals 040931FI
Description
Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis) is used as an electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Golden | |
| 7440-57-5 | |
| 50 x 50 mm | |
| Odorless | |
| Wire Cloth | |
| MFCD00003436 | |
| Insoluble in water. | |
| [Au] | |
| 196.97 | |
| CHEBI:30050 |
| Gold | |
| Gold colored gauze | |
| 100 Mesh | |
| 99.99% (Metals basis) | |
| Au | |
| colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| gold | |
| 23985 |
Chemical Identifiers
| 7440-57-5 | |
| 196.97 | |
| PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
| 23985 | |
| gold |
| Au | |
| MFCD00003436 | |
| colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
| CHEBI:30050 | |
| [Au] |
Safety and Handling
EINECSNumber : 231-165-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only