missing translation for 'onlineSavingsMsg'
Learn More

Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis)

Catalog No. AA40931FF
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
50 x 100 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40931FF 25 x 25 mm
AA40931FI 50 x 50 mm
AA40931FY 50 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40931FF Supplier Thermo Scientific Chemicals Supplier No. 040931FF

Form: Wire cloth 100 x 100mm Approx.4.9g; Open area: 56.3%; Width of opening: 0.0075 in. | CAS: 7440-57-5 | Au | 196.97 g/mol

Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis) is used as an electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Golden
Appearance Gold colored gauze
Density 19.32g/cm3 at 20°C
Quantity 25 x 25 mm
Mesh Size 100 Mesh
Odor Odorless
Form Wire Cloth
Assay Percent Range 99.99% (Metals basis)
Solubility Information Insoluble in water.
Chemical Name or Material Gold
EINECSNumber 231-165-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only