missing translation for 'onlineSavingsMsg'
Learn More

Gold foil, 0.5mm (0.02in) thick, Premion™, 99.999% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol

$2993.96 - $2993.96

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AA44164FF
View Documents
Thermo Scientific Chemicals
44164FF
25 x 25 mm
Each for $2,993.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]

Specifications

7440-57-5
Odorless
Foil
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
99.999%
Gold foil, 0.5mm (0.02 in.) thick
25 x 25 mm
≈6g/25x25mm
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
196.97
CHEBI:30050
Premion
(metals basis)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-165-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only