missing translation for 'onlineSavingsMsg'
Learn More

Gold flake/sphere, APS 1.5-3.0 micron, 99.96% (metals basis), Thermo Scientific™

Catalog No. p-7051874
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4215403 1 g
AA4215406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4215403 Supplier Thermo Scientific Chemicals Supplier No. 4215403
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Quantity 1 g
Particle Size APS 1.5 to 3.0 Micron
Form Flake
Percent Purity 99.96%
Assay (metals basis)
Chemical Name or Material Gold flake/sphere
EINECSNumber 231-165-9
TSCA Yes
Recommended Storage Ambient temperatures