missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold Chloride, Trihydrate, ACS, 49%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor G10255GM
Specifications
| 16961-25-4 | |
| 5 g | |
| VDLSFRRYNGEBEJ-UHFFFAOYSA-K | |
| trichlorogold hydrochloride | |
| 49% | |
| Amber Glass Bottle |
| 1 | |
| AuCl4H | |
| Cl.Cl[Au](Cl)Cl | |
| 339.77 | |
| ACS |
Chemical Identifiers
| 16961-25-4 | |
| 339.77 | |
| trichlorogold hydrochloride |
| AuCl4H | |
| VDLSFRRYNGEBEJ-UHFFFAOYSA-K | |
| Cl.Cl[Au](Cl)Cl |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.