missing translation for 'onlineSavingsMsg'
Learn More

Gold Chloride, Trihydrate, ACS, 49%, Spectrum™ Chemical

Catalog No. p-7165599
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610751 1 g Amber Glass Bottle
18610752 5 g Amber Glass Bottle
18610753 10 g Amber Glass Bottle
18610754 25 g Amber Glass Bottle
18610755 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610751 Supplier Spectrum Chemical Mfg Cor Supplier No. G10251GM
Only null left

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

CAS Max % 1
Quantity 1 g
Percent Purity 49%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.