missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold Chloride, 1% (w/v), Ricca Chemical
Toner For Gomori Methenamine Silver Nitrate Stain
$2,471.84 - $4,738.48
Chemical Identifiers
| CAS | 16961-25-4 , 7732-18-5 |
|---|---|
| Molecular Formula | AuCl4H |
| Molecular Weight (g/mol) | 339.77 |
| InChI Key | VDLSFRRYNGEBEJ-UHFFFAOYSA-K |
| IUPAC Name | trichlorogold hydrochloride |
| SMILES | Cl.Cl[Au](Cl)Cl |
Chemical Identifiers
| 16961-25-4 , 7732-18-5 | |
| 339.77 | |
| trichlorogold hydrochloride |
| AuCl4H | |
| VDLSFRRYNGEBEJ-UHFFFAOYSA-K | |
| Cl.Cl[Au](Cl)Cl |
Specifications
| 16961-25-4 , 7732-18-5 | |
| 100.0, 1.01 | |
| AuCl4H | |
| Cl.Cl[Au](Cl)Cl | |
| 339.77 | |
| Laboratory | |
| Yellow | |
| Gold Chloride |
| 97.0, 0.97 | |
| 1% (w/v) | |
| VDLSFRRYNGEBEJ-UHFFFAOYSA-K | |
| trichlorogold hydrochloride | |
| 1% W/V | |
| Aqueous Solution | |
| Liquid |