missing translation for 'onlineSavingsMsg'
Learn More

Gold, AAS standard solution, Specpure™ Au 1000μg/mL

Catalog No. p-7080411
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA88068AE 100 mL
AA88068AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA88068AE Supplier Thermo Scientific Chemicals Supplier No. 088068AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gold, AAS standard solution is used for the determination of gold by flame atomic absorption spectrometry (FAAS) and graphite furnace atomic absorption spectrometry (GFAAS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00003436

Specifications

Physical Form Liquid
Concentration Au in 20% HCl
UN Number UN1789
Solubility Miscible with water
Quantity 100 mL
Grade Specpure
Chemical Name or Material Gold, AAS standard solution, Au 1000μg/mL
Hazard Category H290-H314-H335
Hazard Statement GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only