missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Goat Serum
Description
This serum is produced from whole blood collected from healthy adult animals.
Specifications
Specifications
| Color | Reddish brown |
| Content And Storage | Store at -20°C |
| Form | Liquid |
| Quantity | 100 mL |
| Sterilization Method | 0.2 μm filtration |
| Source | Whole blood collected from healthy adult animals |
| For Use With (Application) | Cell Culture |
| pH | 7 to 8 |