missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat IgG anti-Guinea Pig Complement C3, Polyclonal Antibody, FITC, MP Biomedicals
Supplier: MP Biomedicals Inc 0855385
Description
Host Species: Goat
Species Reactivity: Gp
IFA, Cell Staining, Tissue Staining,Western Blotting, ACIF Assays
Specifications
| Guinea Pig Complement C3 | |
| FITC | |
| 2 mL | |
| Guinea Pig |
| Polyclonal | |
| Goat | |
| Primary | |
| Lyophilized |