missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab) Fragment to Hu Lambda Chain (bound), FITC, Polyclonal Secondary Antibody, MP Biomedicals
Description
Host Species: Goat
Species Reactivity: Hu
Immunofluorescence, Fluorescence Microscopy, Fluorescent Activated Cell Sorting, Western Blotting
Specifications
Specifications
| Antigen | IgG |
| Classification | Polyclonal |
| Conjugate | FITC |
| Host Species | Goat |
| Quantity | 2 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Target Species | Human |
| Form | Purified |