missing translation for 'onlineSavingsMsg'
Learn More

Light chain Goat anti-Rat, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP202CMI
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP202CMI 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP202CMI Supplier MilliporeSigma Supplier No. AP202C
Only null left

Goat Polyclonal Antibody

Detect light chain using this Goat anti-Rat light chain Antibody, Cy3 conjugate validated for use in ELISA and western blotting.

Specifications

Antigen Goat anti-Rat light chain
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Cyanine3
Formulation Purified by immunoaffinity chromatography. Liquid in 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 0.05% Sodium Azide. 15mg/mL BSA as stabilizer.
Host Species Goat
Immunogen purified rat IgG light chain.
Purification Method Affinity Purified
Quantity 0.5 mg
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C Lyophilized for up to 12 months from date of receipt −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Protect from light
Isotype IgG
Show More Show Less

For Research Use Only