missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rat IgG2b Recombinant Secondary Antibody, PE

Catalog No. 5000153531
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5000153531 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5000153531 Supplier Invitrogen™ Supplier No. SA510377PE

Goat Recombinant Monoclonal Secondary Antibody

Goat anti-Rat IgG2b, Recombinant Secondary Antibody specifically detects the Fc portion of rat IgG2b isotype and shows strong reactivity to total Rat IgG. It has minimal to no cross-reactivity towards other rat IgG isotypes and immunoglobulins tested from other species. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. R-phycoerythrin (R-PE) is an intensely bright phycobiliprotein isolated from red algae that exhibits extremely bright red-orange fluorescence with high quantum yields. In practical applications, the sensitivity of R-PE conjugates is usually 5 to 10 times greater than those of the corresponding fluorescein conjugates. R-PE is a large molecule used for fluorescence-based detection, primarily in flow cytometry, microarray assays, ELISAs, and other applications that require high sensitivity, but not photostability. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rat IgG2b
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 10H38L66
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 5mM sodium azide; pH 7.4
Host Species Goat
Immunogen Purified Rat IgG.
Purification Method Affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less