missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG, Whole Molecule Polyclonal Secondary Antibody, HRP, MP Biomedicals
Supplier: MP Biomedicals Inc 0855741
Description
Host Species: Goat
Species Reactivity: Rat
Specifications
| IgG, Whole Molecule | |
| HRP | |
| 2 mg | |
| Rat |
| Polyclonal | |
| Goat | |
| Secondary | |
| Purified |