missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Rat, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Goat anti-Rat IgG Antibody, HRP conjugate, Species Adsorbed detects level of Rat IgG and has been published and validated for use in ELISA and western blotting.
Specifications
Specifications
| Antigen | Goat anti-Rat IgG Species adsorbed |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.8mg/mL |
| Conjugate | HRP |
| Formulation | Lyophilized. Buffer = 0.01M sodium phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA. Contains no preservative. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water. |
| Host Species | Goat |
| Quantity | 500 μL |
| Primary or Secondary | Secondary |
| Target Species | Rat |
| Show More |
For Research Use Only